Learn More
Invitrogen™ NPR3 Recombinant Rabbit Monoclonal Antibody (24GB3585)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557098
 View more versions of this product
Product Code. 30275662
  • £126.00 / 20µL
Description
NPR3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NPR3
Recombinant Monoclonal
1.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P17342, P41740, P70180
NPR3
A synthesized peptide derived from human Natriuretic Peptide Receptor C (aa 500-541).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3585
Unconjugated
NPR3
A NPR-C; ANP-C; ANP-CR; ANPRC; ANPR-C; atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; atrial natriuretic peptide C-type receptor; atrial natriuretic peptide receptor 3; atrial natriuretic peptide receptor 3-like; atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; C5orf23; EF-2; guanylate cyclase C; GUCY2B; lgj; longjohn; natriuretic peptide receptor 3; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; Npr3; NPRC; NPR-C; stri; strigosus
Rabbit
Affinity chromatography
RUO
18162, 25339, 4883
-20°C