missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599836
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NPR2.
This gene encodes natriuretic peptide receptor B, one of two integral membrane receptors for natriuretic peptides. Both NPR1 and NPR2 contain five functional domains: an extracellular ligand-binding domain, a single membrane-spanning region, and intracellularly a protein kinase homology domain, a helical hinge region involved in oligomerization, and a carboxyl-terminal guanylyl cyclase catalytic domain. The protein is the primary receptor for C-type natriuretic peptide, which upon ligand binding exhibits greatly increased guanylyl cyclase activity. Mutations in this gene are the cause of acromesomelic dysplasia Maroteaux type.
Specifications
| NPR2 | |
| Polyclonal | |
| Unconjugated | |
| Npr2 | |
| AMDM; ANPb; ANP-B; ANPRB; ANPR-B; atrial natriuretic peptide B-type receptor; atrial natriuretic peptide receptor 2; atrial natriuretic peptide receptor B; atrial natriuretic peptide receptor type B; cn; ECDM; GC-B; GC-B2; GC-B3; guanylate cyclase 2B; guanylate cyclase B; guanylyl cyclase-B; GUC2B; GUCY2B; mNPR-B; natriuretic peptide receptor 2; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); natriuretic peptide receptor-B; Nppb receptor; NPR2; NPRB; NPR-B; NPRBi; pwe; SNSK | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116564, 230103, 4882 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16067, P20594, Q6VVW5 | |
| Npr2 | |
| A synthesized peptide derived from human NPR2(Accession P20594), corresponding to amino acid residues S647-L697. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction