missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NPPB
GREENER_CHOICE

Product Code. 18064932
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18064932 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18064932 Supplier Tocris Bioscience™ Supplier No. 0593/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits calcium-sensitive chloride currents

Inhibits calcium-sensitive chloride currents (10 μM). Putative GPR35 agonist.
TRUSTED_SUSTAINABILITY

Specifications

CAS 107254-86-4
Quantity 50 mg
Molecular Formula C16H16N2O4
Formula Weight 300.31
Purity >99%
Molecular Weight (g/mol) 300.31
Chemical Name or Material NPPB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.