missing translation for 'onlineSavingsMsg'
Learn More

NPPB, MedChemExpress

Product Code. 16489776
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489776 50 mg 50mg
16474496 100 mg 100mg
16403238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489776 Supplier Medchem Express Supplier No. HY101012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NPPB is a blocker of the outwardly rectifying chloride channel (ORCC).

NPPB is a blocker of the outwardly rectifying chloride channel (ORCC).

Specifications

Chemical Name or Material NPPB
Physical Form Solid
CAS 107254-86-4
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H16N2O4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 150 mg/mL (499.48 mM; Need ultrasonic and warming)
SMILES O=C(O)C1=CC([N+]([O-])=O)=CC=C1NCCCC2=CC=CC=C2
Molecular Weight (g/mol) 300.31
Formula Weight 300.31
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.