Learn More
Invitrogen™ NPPB/BNP Human ELISA Kit
Sandwich ELISA Kit
£546.00 - £2320.00
Specifications
Accession Number | P16860 |
---|---|
Assay Sensitivity | 14 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15475083
|
Invitrogen™
EHNPPB |
96 Tests |
£546.00
1 set |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
17105529
|
Invitrogen™
EHNPPBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
Description
The Human NPPB/BNP ELISA quantitates Hu NPPB in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NPPB. Principle of the method The Human NPPB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
In cardiac tissue brain natriuretic peptide (BNP) is synthesized as 134 amino acid precursor (prepro-BNP), which is cleaved by proteases to form a 26 aa ""signal"" peptide and a 108 aa pro-BNP. Proteolytic digestion of pro-BNP results in formation of 76 aa amino-terminal NT-proBNP and biologically active 32 aa BNP hormone molecule. Both proBNP and NTproBNP circulate in human plasma and have been proposed as markers for early diagnosis of left ventricular dysfunction as well as prognostic markers of possible cardiac complications at patients with heart failure.Specifications
P16860 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
BNP | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
14 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA pair antibody detects human BNP. Other species not determined. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
4879 | |
<12% | |
HRP | |
Angiogenesis | |
BNP | |
1 hr. 20 min. |