missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557602
This item is not returnable.
View return policy
Description
Immunogen sequence: APVTMDSFFF GCELSGHTRS FTFKVEEEDD AEHVLALTML CLTEGAKDEC NVV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.
Specifications
| NPM3 | |
| Polyclonal | |
| Unconjugated | |
| NPM3 | |
| Npm3; Nub1; nucleophosmin/nucleoplasmin 3; nucleophosmin/nucleoplasmin family, member 3; nucleophosmin/nucleoplasmin, 3; nucleoplasmin 3; nucleoplasmin-3; PORMIN; RGD1565542; TMEM123 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75607 | |
| NPM3 | |
| Recombinant protein corresponding to Human NPM3. Recombinant protein control fragment (Product #RP-97051). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction