missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPLOC4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA554749

Product Code. 15712681

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LEPFDEDYLN HLEPPVKHMS FHAYIRKLTG GADKGKFVAL ENISCKIKSG CEGHLPWPNG ICTKCQPSAI TLNRQKYRHV DNIMFENHTV ADRFLDFWRK TGNQHFGYL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

NPLOC4 gene ontology annotations related to this gene include ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NPLOC4
Polyclonal
Unconjugated
NPLOC4
AK129375; FLJ20657; homolog of yeast nuclear protein localization 4; KIAA1499; mKIAA1499; NPL4; NPL4 homolog, ubiquitin recognition factor; Nploc4; NPLOC4 ubiquitin recognition factor; nuclear protein localization 4 homolog; nuclear protein localization 4 homolog (S. cerevisiae); nuclear protein localization protein 4 homolog; Protein NPL4
Rabbit
Antigen affinity chromatography
RUO
55666
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TAT6
NPLOC4
Recombinant protein corresponding to Human NPLOC4. Recombinant protein control fragment (Product #RP-93492).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.