missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113402
This item is not returnable.
View return policy
Description
NPL Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Catalyzes the cleavage of N-acetylneuraminic acid (sialic acid) to form pyruvate and N-acetylmannosamine via a Schiff base intermediate. It prevents sialic acids from being recycled and returning to the cell surface. Involved in the N- glycolylneuraminic acid (Neu5Gc) degradation pathway. Although human is not able to catalyze formation of Neu5Gc due to the inactive CMAHP enzyme, Neu5Gc is present in food and must be degraded.
Specifications
| NPL | |
| Polyclonal | |
| Unconjugated | |
| NPL | |
| 0610033B02Rik; C112; C1orf13; dihydrodipicolinate synthetase homolog 1; N-acetylneuraminate lyase; N-acetylneuraminate pyruvate lyase; N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase); N-acetylneuraminate pyruvate-lyase; N-acetylneuraminic acid aldolase; NAL; NALase; Npl; NPL1; Sialate lyase; sialate-pyruvate lyase; Sialic acid aldolase; sialic acid lyase | |
| Rabbit | |
| Protein G | |
| RUO | |
| 74091, 80896 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BXD5, Q9DCJ9 | |
| NPL | |
| Recombinant Human N-acetylneuraminate lyase protein (17-77AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction