missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPL Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5140278

Product Code. 17894790

  • £387.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FVNGTTGEGL SLSVSERRQV AEEWVTKGKD KLDQVIIHVG ALSLKESQEL AQHAAEIGAD Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 88%.

Catalyzes the cleavage of N-acetylneuraminic acid (sialic acid) to form pyruvate and N-acetylmannosamine via a Schiff base intermediate. It prevents sialic acids from being recycled and returning to the cell surface. Involved in the N- glycolylneuraminic acid (Neu5Gc) degradation pathway. Although human is not able to catalyze formation of Neu5Gc due to the inactive CMAHP enzyme, Neu5Gc is present in food and must be degraded.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NPL
Polyclonal
Unconjugated
NPL
0610033B02Rik; C112; C1orf13; dihydrodipicolinate synthetase homolog 1; N-acetylneuraminate lyase; N-acetylneuraminate pyruvate lyase; N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase); N-acetylneuraminate pyruvate-lyase; N-acetylneuraminic acid aldolase; NAL; NALase; Npl; NPL1; Sialate lyase; sialate-pyruvate lyase; Sialic acid aldolase; sialic acid lyase
Rabbit
Antigen affinity chromatography
RUO
80896
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BXD5
NPL
Recombinant protein corresponding to Human NPL. Recombinant protein control fragment (Product #RP-109500).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.