missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPIPL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | NPIPL2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204143
|
Novus Biologicals
NBP2-59784 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623577
|
Novus Biologicals
NBP2-59784-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NPIPL2 Polyclonal specifically detects NPIPL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NPIPL2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| A-761H5.4, LOC440348, nuclear pore complex interacting protein-like 2, Nuclear Pore Complex-Interacting Protein-Like 2 | |
| NPIPL2 | |
| IgG | |
| Protein A purified |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 440348 | |
| This antibody was developed against a recombinant protein corresponding to the amino acid sequence:EQSPKPKRQREAEAQQLPKPKRRRLSKLRTRHCTQAWAIRINP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title