missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPHP5/IQCB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15747-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| NPHP5/IQCB1 | |
| Polyclonal | |
| Unconjugated | |
| IQCB1 | |
| IQ motif containing B1, IQCB1, KIAA0036, Nephrocystin 5, NPHP5, PIQ, SLSN5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303915, 320299, 9657 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15051, Q8BP00 | |
| Iqcb1 | |
| NPHP5,IQCB1 Fusion Protein Ag8203 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction