missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPHP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539283
This item is not returnable.
View return policy
Description
This gene encodes a protein involved in renal tubular development and function. This protein interacts with nephrocystin, and belongs to a multifunctional complex that is localized to actin- and microtubule-based structures. Mutations in this gene are associated with nephronophthisis type 4, a renal disease, and with Senior-Loken syndrome type 4, a combination of nephronophthisis and retinitis pigmentosa. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Apr 2014].
Specifications
| NPHP4 | |
| Polyclonal | |
| Unconjugated | |
| NPHP4 | |
| KIAA0673 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 261734 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75161 | |
| NPHP4 | |
| A synthetic peptide derived from the internal region of human NPHP4 (CRQGKGPQDVSRESD) | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction