missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPHP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564227
This item is not returnable.
View return policy
Description
Immunogen sequence: SQALVLRSRL RLPEMVGHPA FAVIFQLEYV FSSPAGVDGN AASVTSLSNL ACMHMVRWAV WNPLLEADSG RVTLPLQGGI QPNPSHCLVY KVPSASMSSE EVKQVESGTL RFQ Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
This gene encodes a protein involved in renal tubular development and function. This protein interacts with nephrocystin, and belongs to a multifunctional complex that is localized to actin- and microtubule-based structures. Mutations in this gene are associated with nephronophthisis type 4, a renal disease, and with Senior-Loken syndrome type 4, a combination of nephronophthisis and retinitis pigmentosa. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Apr 2014].
Specifications
| NPHP4 | |
| Polyclonal | |
| Unconjugated | |
| NPHP4 | |
| KIAA0673 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 261734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75161 | |
| NPHP4 | |
| Recombit protein corresponding to Human NPHP4 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction