missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPFF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533982
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gibbon (95%) Gorilla (85%) Monkey, Elephant (80%).
The central nervous system neuropeptide FF (NPFF), is believed to play a role in pain modulation and opiate tolerance as well as other physiological functions. Two G protein-coupled receptors, NPFF1 and NPFF2, were isolated from the human central nervous system.
Specifications
| NPFF1 | |
| Polyclonal | |
| Unconjugated | |
| NPFFR1 | |
| ff1; FLJ10751; G protein-coupled receptor 147; Gm236; Gpr147; G-protein coupled receptor 147; G-protein coupled receptor NPFF1R; neuropeptide FF 1; neuropeptide FF receptor 1; NPFF1; NPFF1 receptor; NPFF1R1; NPFFR1; NPFF-R1; OT7T022; RFamide-related peptide receptor; RFamide-related peptide receptor OT7T022 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64106 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9GZQ6 | |
| NPFFR1 | |
| Synthetic 20 amino acid peptide from C-terminus of human NPFF1 Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction