missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPFF Polyclonal Antibody

Brand: Thermo Scientific PA5115244
This item is not returnable.
View return policy
Description
Specifications
| NPFF | |
| Polyclonal | |
| Unconjugated | |
| NPFF | |
| FMRFAL; FMRFamide-related peptides; Neuropeptide AF; Neuropeptide AF-like; Neuropeptide FF; neuropeptide FF-amide peptide precursor; Neuropeptide SF; NPAF; Npff; NPSF; Pro-FMRFamide-related neuropeptide FF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8620 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O15130 | |
| NPFF | |
| A synthesized peptide derived from human NPFF(Accession O15130), corresponding to amino acid residues Q58-Q108. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction