missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPEPPS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522383
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (97%), Rat (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the puromycin-sensitive aminopeptidase, a zinc metallopeptidase which hydrolyzes amino acids from the N-terminus of its substrate. The protein has been localized to both the cytoplasm and to cellular membranes. This enzyme degrades enkaphalins in the brain, and studies in mouse suggest that it is involved in proteolytic events regulating the cell cycle.
Specifications
| NPEPPS | |
| Polyclonal | |
| Unconjugated | |
| NPEPPS | |
| AAP-S; aminopeptidase puromycin sensitive; cytosol alanyl aminopeptidase; goku; metalloproteinase MP100; MP100; NPEPPS; Psa; Puromycin-sensitive aminopeptidase; R74825 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19155, 50558, 9520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.68 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P55786, Q11011 | |
| NPEPPS | |
| Recombinant protein encompassing a sequence within the center region of human NPEPPS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction