missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPC1L1 Polyclonal Antibody
Description
The target sequence has 93% sequence homology with human.
Dietary consumption and intestinal cholesterol absorption contribute to plasma cholesterol levels, which factor into the risk of coronary heart disease. Niemann-Pick type C1 Like 1 protein (NPC1L1 or NPC3) is enriched in the small intestine and is found in the brush border membrane of enterocytes. It plays a critical role in absorption of intestinal cholesterol.
Specifications
Specifications
| Antigen | NPC1L1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate, tris citrate with 0.1% sodium azide; pH 7-8 |
| Gene | Npc1l1 |
| Gene Accession No. | Q6T3U3, Q6T3U4, Q9UHC9 |
| Gene Alias | 9130221N23Rik; BC 400-127; Gm243; Niemann-Pick C1-like protein 1; Niemann-Pick C3; Niemann-Pick disease, type C1; Niemann-Pick disease, type C1-like 1; NPC1 (Niemann-Pick disease, type C1, gene)-like 1; NPC1 like 1; NPC1 like intracellular cholesterol transporter 1; NPC11L1; NPC1L1; NPC1-like 1; NPC1-like intracellular cholesterol transporter 1; NPC3 |
| Gene Symbols | Npc1l1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?