Learn More
Invitrogen™ NPC1 Recombinant Rabbit Monoclonal Antibody (24GB620)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557129
 View more versions of this product
Product Code. 30276517
  • £434.00 / 100µL
Description
NPC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Mutations in the Niemann-Pick disease type C1 (NPC1) gene result in a fatal progressive neurodegenerative disorder characterized by an abnormal sequestration of lipids including cholesterol and glycosphingolipids. The NPC1 protein is a large protein that resides in the limiting membrane of endosomes and lysosomes and mediates intracellular cholesterol trafficking via binding of cholesterol to its N-terminal domain. NPC1 transports low-density lipoproteins to late endosomal/lysosomal compartments where they are hydrolyzed and released as free cholesterol. NPC1, in addition to FTO, MC4R, and PTER has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NPC1
Recombinant Monoclonal
0.85 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O15118, O35604
NPC1
A synthesized peptide derived from human Niemann Pick C1 (aa 1240-1278).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB620
Unconjugated
NPC1
A430089E03Rik; C85354; Cdig2; D18Ertd139e; D18Ertd723e; FLJ98532; I79_014078; lcsd; Niemann Pick type C1; Niemann-Pick C disease protein; niemann-Pick C1 protein; Niemann-Pick disease, type C1; Niemann-Pick type C1; Niemann-Pick type C1 protein; nmf164; NPC; NPC intracellular cholesterol transporter 1; NPC1; sphingomyelinosis; spm; truncated Niemann-Pick C1
Rabbit
Affinity chromatography
RUO
18145, 266732, 4864
-20°C