missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPBWR1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | NPBWR1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NPBWR1 Polyclonal antibody specifically detects NPBWR1 in Human samples. It is validated for Western BlotSpecifications
| NPBWR1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.3), 50% glycerol | |
| 2831 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| G protein-coupled receptor 7, GPR7 neuropeptides B/W receptor type 1, G-protein coupled receptor 7, MGC129755, neuropeptides B/W receptor 1, opioid-somatostatin-like receptor 7 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human NPBWR1 (NP_005276.2). RTYSAARAVSLAVWGIVTLVVLPFAVFARLDDEQGRRQCVLVFPQPEAFWWRASRLYTLVLGFAIPVSTICVLYTTLLCRLHAMRLDSHAKALERAKKRVT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title