missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPAS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598603
This item is not returnable.
View return policy
Description
NPAS4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
Specifications
| NPAS4 | |
| Polyclonal | |
| Unconjugated | |
| NPAS4 | |
| bHLH PAS type transcription factor NXF; bHLHe79; bHLH-PAS type transcription factor NXF; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; neuronal PAS domain protein 4; neuronal PAS domain-containing protein 4; Neuronal PAS4; Npas4; NXF; PAS domain-containing protein 10; PASD10; PASD10 and LE-PAS | |
| Rabbit | |
| Protein G | |
| RUO | |
| 266743 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IUM7 | |
| NPAS4 | |
| Recombinant Human Neuronal PAS domain-containing protein 4 protein (445-585AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction