missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPAS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540561
This item is not returnable.
View return policy
Description
Peptide sequence: QAHLDSPSQT FPEQLSPNPT KTYFAQEGCS FLYEKLPPSP SSPGNGDCTL Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 86%.
NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
Specifications
| NPAS4 | |
| Polyclonal | |
| Unconjugated | |
| NPAS4 | |
| bHLH PAS type transcription factor NXF; bHLHe79; bHLH-PAS type transcription factor NXF; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; neuronal PAS domain protein 4; neuronal PAS domain-containing protein 4; Neuronal PAS4; Npas4; NXF; PAS domain-containing protein 10; PASD10; PASD10 and LE-PAS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 266743 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IUM7 | |
| NPAS4 | |
| Synthetic peptide directed towards the middle region of human NPAS4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction