missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Npas4 Mouse anti-Rat, HRP, Clone:S408-79, Abnova™
Mouse monoclonal antibody raised against partial recombinant rat Npas4.
Brand: Abnova MAB17369.100ug
This item is not returnable.
View return policy
Specifications
| Npas4 | |
| Monoclonal | |
| HRP | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Npas4 | |
| Mouse | |
| 100 μg | |
| Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S408-79 | |
| neuronal PAS domain protein 4 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Nxf | |
| Recombinant protein corresponding to amino acids 597-802 at C-terminus of rat Npas4. | |
| 266734 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction