Learn More
Npas4 Antibody - Unpurified, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-06574
This item is not returnable.
View return policy
Description
Npas4 Polyclonal specifically detects Npas4 in Mouse, Rat, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:100-1:2000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| NPAS4 | |
| Unpurified | |
| Mouse, Rat |
| LYOPH | |
| BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10 | |
| Synthetic peptide derived from Cterm domain of mouse NPAS4 protein | |
| RUO | |
| Antisera |