missing translation for 'onlineSavingsMsg'
Learn More

NPAS3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18382127 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382127 100 μg 100µL
18387452 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18382127 Supplier Novus Biologicals Supplier No. NBP316051100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPAS3 Polyclonal antibody specifically detects NPAS3 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAS3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias Basic-helix-loop-helix-PAS protein MOP6, BHLHE12, bHLHe12FLJ11605, Class E basic helix-loop-helix protein 12, Member of PAS protein 6, MOP6FLJ10003, neuronal PAS domain protein 3, neuronal PAS domain-containing protein 3, Neuronal PAS3, PAS domain-containing protein 6, PASD6FLJ11138
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human NPAS3 (NP_001158221.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 64067
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.