missing translation for 'onlineSavingsMsg'
Learn More

NPAS3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18422153 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422153 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422153 Supplier Novus Biologicals Supplier No. NBP176785AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPAS3 Polyclonal antibody specifically detects NPAS3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAS3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Basic-helix-loop-helix-PAS protein MOP6, BHLHE12, bHLHe12FLJ11605, Class E basic helix-loop-helix protein 12, Member of PAS protein 6, MOP6FLJ10003, neuronal PAS domain protein 3, neuronal PAS domain-containing protein 3, Neuronal PAS3, PAS domain-containing protein 6, PASD6FLJ11138
Host Species Rabbit
Immunogen Antibody was raised against a 28 amino acid synthetic peptide from near the amino terminus of human NPAS3. The immunogen is located within amino acids 40 - 90 of NPAS3. Amino Acid Squence: QALRKEKSRDAARSRRGKENFEFYELAK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 64067
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.