missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPAS1 Polyclonal Antibody
Description
Positive Control: U87-MG, IMR32, SK-N-AS Predicted Reactivity: Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.
Specifications
Specifications
| Antigen | NPAS1 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | NPAS1 |
| Gene Accession No. | P97459, Q99742 |
| Gene Alias | basic-helix-loop-helix-PAS protein MOP5; BHLHE11; Class E basic helix-loop-helix protein 11; member of PAS protein 5; member of PAS superfamily 5; MOP5; neuronal PAS domain protein 1; neuronal PAS domain-containing protein 1; Neuronal PAS1; Npas1; PAS domain-containing protein 5; PASD5 |
| Gene Symbols | NPAS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?