missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPAS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567232
This item is not returnable.
View return policy
Description
Immunogen sequence: VLWVSHVLSQA EGGQTPLDAF QLPASVACEE ASSPGPEPTE PEPPTEGKQA APAENEAPQT QGQRIKV Highest antigen sequence identity to the following orthologs - mouse 70%, rat 70%.
NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.
Specifications
| NPAS1 | |
| Polyclonal | |
| Unconjugated | |
| NPAS1 | |
| basic-helix-loop-helix-PAS protein MOP5; BHLHE11; Class E basic helix-loop-helix protein 11; member of PAS protein 5; member of PAS superfamily 5; MOP5; neuronal PAS domain protein 1; neuronal PAS domain-containing protein 1; Neuronal PAS1; Npas1; PAS domain-containing protein 5; PASD5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4861 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99742 | |
| NPAS1 | |
| Recombinant Human NPAS1. Recombinant protein control fragment (Product #RP-107868). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction