Learn More
Socorex™ Nozzle/Adapter for 5mL Straw Tips
Allows you to use 5mL polypropylene straw tips with an Acura™ Manual 835 adjustable volume macropipette. Socorex™ Nozzle/Adapter for 5mL Straw Tips lets you use straw tips to reach into deep, narrow vials and Stomacher bags.
Brand:  Socorex™ 1.831.707
Additional Details : Weight : 0.01100kg
Product Code. 15740639
  • £16.75 / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • Fits Acura™ Manual 835 adjustable volume macropipette.
  • Accepts 5mL volume straw tips.
  • Facilitates reach into deep, narrow vials and Stomacher bags.
  • Facilitates pickup of particle-containing liquids.
Specifications
Specifications
Acura™ 835 Manual adjustable volume macropipette, 5mL straw tips
1/ea.