missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOXA2/p67phox Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67594-1-IG-20UL
This item is not returnable.
View return policy
Description
The NOXA2/p67phox antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOXA2/p67phox. This antibody recognizes human, pig antigen. The NOXA2/p67phox antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| NOXA2/p67phox | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ93058; NCF-2; NOXA2; P67-PHOX; P67PHOX | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| 67 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1D11A3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 4688 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction