missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOXA1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94716-0.1ml
This item is not returnable.
View return policy
Description
NOXA1 Polyclonal antibody specifically detects NOXA1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NOXA1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| Antigen NY-CO-31, FLJ25475, inhibitory NADPH oxidase activator 1, MGC131800, NADPH oxidase activator 1, NCF2-like protein, NOX activator 1, NY-CO-31, p51-nox, p51NOX, P67phox-like factor, SDCCAG31, serologically defined colon cancer antigen 31 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-300 of human NOXA1 (NP_006638.1). EAASSLREAMSKWPEGSLNGLDSALDQVQRRGSLPPRQVPRGEVFRPHRWHLKHLEPVDFLGKAKVVASAIPDDQGWGVRPQQPQGPGANHDARSLIMDSPRAGTHQGPLDAETEVGADRCTSTAYQEQRPQVEQVGKQAPLSPGLPAMGGPGPGPCEDPA | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
| 10811 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction