Learn More
NOXA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA118331
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
Apoptosis is related to many diseases and development. The p53 tumor-suppressor protein induces apoptosis through transcriptional activation of several genes including p53R2, p53AIP1, and PUMA. A new p53 target gene, Noxa, was recently identified, which encodes a protein belonging to the subfamily of BH3-only proapoptic proteins. Noxa and PUMA are both transcriptional targets of p53 and BH3-only proteins. X-ray irradiation increased p53-dependent Noxa mRNA and protein levels. Noxa, when ectopically expressed, interacted with anti-apoptotic Bcl-2 family members, resulting in the activation of caspase-9. Noxa, like PUMA, localized to mitochondria and induces apoptosis in response to p53. Noxa and PUMA may represent direct mediators of p53-induced apoptosis. Increased levels of p53 and its target gene Noxa was found in the impaired tumor development.Specifications
NOXA | |
Polyclonal | |
Unconjugated | |
PMAIP1 | |
adult T cell leukemia-derived PMA-responsive; APR; Immediate-early-response protein APR; Noxa; Phorbol-12-myristate-13-acetate-induced protein 1; PMA-ind; PMA-induced protein 1; Pmaip1; Protein Noxa | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
Q9JM54 | |
PMAIP1 | |
Synthetic peptide corresponding to the N-terminal residues M(1) P G R K A R R N A P V N P T R(16) of mouse Noxa. | |
RUO | |
58801 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |