missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOXA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141010
This item is not returnable.
View return policy
Description
Suggested positive control: human, mouse or rat thymus tissue lysate or protein, antigen standard for PMAIP1 (transient overexpression lysate). Immunogen sequence homology: amino acids 51-66: Rat (77%); amino acids 75-90: Rat (93%), Human (76%).
Apoptosis is related to many diseases and development. The p53 tumor-suppressor protein induces apoptosis through transcriptional activation of several genes including p53R2, p53AIP1, and PUMA. A new p53 target gene, Noxa, was recently identified, which encodes a protein belonging to the subfamily of BH3-only proapoptic proteins. Noxa and PUMA are both transcriptional targets of p53 and BH3-only proteins. X-ray irradiation increased p53-dependent Noxa mRNA and protein levels. Noxa, when ectopically expressed, interacted with anti-apoptotic Bcl-2 family members, resulting in the activation of caspase-9. Noxa, like PUMA, localized to mitochondria and induces apoptosis in response to p53. Noxa and PUMA may represent direct mediators of p53-induced apoptosis. Increased levels of p53 and its target gene Noxa was found in the impaired tumor development.
Specifications
| NOXA | |
| Polyclonal | |
| Unconjugated | |
| PMAIP1 | |
| adult T cell leukemia-derived PMA-responsive; APR; Immediate-early-response protein APR; Noxa; Phorbol-12-myristate-13-acetate-induced protein 1; PMA-ind; PMA-induced protein 1; Pmaip1; Protein Noxa | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5366, 58801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q13794, Q9JM54 | |
| PMAIP1 | |
| Mixture of synthetic peptides containing amino acids 51-66 and 75-90 of mouse Noxa. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction