missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Noxa Monoclonal antibody specifically detects Noxa in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Noxa |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 69N15C9 |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa |
| Host Species | Rat |
| Immunogen | This Noxa antibody (69N15C9) is made against amino acids 1-103 of mouse Noxa. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?