missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (114C307.1) - Non-Recombinant Monoclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Noxa Monoclonal specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Noxa |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 114C307.1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1 ug/ million cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunohistochemistry-Paraffin 5 ug/ml |
| Gene Accession No. | Q13794 |
| Gene Alias | adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa |
| Gene Symbols | PMAIP1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?