missing translation for 'onlineSavingsMsg'
Learn More

Noxa Antibody (114C307.1), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18420344 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420344 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420344 Supplier Novus Biologicals Supplier No. NB6001159MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Noxa Monoclonal antibody specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Noxa
Applications Flow Cytometry
Classification Monoclonal
Clone 114C307.1
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Host Species Mouse
Immunogen This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5366
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.