missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580860
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NOX4.
Oxygen sensing is essential for homeostasis in all aerobic organisms. A phagocyte-type oxidase, similar to that responsible for the production of large amounts of reactive oxygen species (ROS) in neutrophil granulocytes, with resultant antimicrobial activity, has been postulated to function in the kidney as an oxygen sensor that regulates the synthesis of erythropoietin in the renal cortex. NOX4 has a role as a redox messenger in the activation of intracellular signaling pathways leading (or contributing) to mitochondriogenesis, cell survival, and differentiation in hematopoietic stem cells. Data suggests that NOX4 provides a novel link between the insulin receptor and the generation of cellular reactive oxygen species that enhances insulin signal transduction.
Specifications
| NOX4 | |
| Polyclonal | |
| Unconjugated | |
| Nox4 | |
| AI648021; kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX1; KOX-1; NADPH oxidase 4; NOX 4; NOX4; NOX-4; renal NAD(P)H-oxidase; RENOX; superoxide-generating NADPH oxidase 4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 50507 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9NPH5 | |
| Nox4 | |
| A synthetic peptide corresponding to the C-terminus of the Human NOX4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction