missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nox4 Antibody (NOX4/1245), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54338AF532
This item is not returnable.
View return policy
Description
Nox4 Monoclonal specifically detects Nox4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Nox4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NOX4 | |
| Recombinant human Nox4 protein (Uniprot: Q9NPH5) | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| NOX4/1245 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| 50507.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction