missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA538036
This item is not returnable.
View return policy
Description
PA5-38036 detects Nox3 in mouse bone marrow and humanG-CSF mobilized CD34+ cells in immunofluorescence analysis. A recommended positive control is 293 cell lysate. NOX3 is not expected to cross-react with other NOX proteins.
The NOX family of NAPDH oxidases is comprised of seven transmembrane proteins that oxidize intracellular NAPDH/NADH, causing electron transport across the membrane and the reduction of molecular oxygen to superoxide. NOX3 is expressed predominantly in the inner ear and is involved in the biogenesis of otoconia/otolith. It has been suggested that NOX3 is activated by the Transient Receptor Potential Vanilloid 1 (TRPV1), and this activity causes increased levels of reactive oxygen species in the inner ear, which in turn leads to STAT1-mediated inflammation and hearing loss.
Specifications
| NOX3 | |
| Polyclonal | |
| Unconjugated | |
| NOX3 | |
| GP91-3; gp91phox homolog 3; head-tilt; het; Mitogenic oxidase 2; MOX2; MOX-2; NADPH oxidase 3; NADPH oxidase catalytic subunit-like 3; NAPDH oxidase 3; nmf250; Nox3; NOX-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 224480, 50508 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q672J9, Q9HBY0 | |
| NOX3 | |
| 15 amino acid peptide near the amino terminus of human NOX3. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction