missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOX2 Recombinant Rabbit Monoclonal Antibody (23GB5135), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | NOX2 |
|---|---|
| Clone | 23GB5135 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088526
![]() |
Invitrogen™
MA552800 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087846
|
Invitrogen™
MA552801 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOX2 Recombinant Monoclonal antibody specifically detects NOX2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| NOX2 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P04839, Q61093 | |
| 13058, 1536, 66021 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5135 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AMCBX2; C88302; Cgd; CGD91-phox; Cybb; Cyd; cytochrome b(558) subunit beta; cytochrome b-245 beta chain; cytochrome b-245 beta polypeptide; cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; cytochrome b558 subunit beta; endothelial type gp91-phox; GP91 PHOX; GP911; GP91-1; GP91PHOX; gp91-phox; Heme-binding membrane glycoprotein gp91phox; IMD34; NADPH oxidase 2; neutrophil cytochrome b 91 kDa polypeptide; Nox2; p22 phagocyte B-cytochrome; p91PHOX; p91-PHOX; superoxide-generating NADPH oxidase heavy chain subunit | |
| A synthesized peptide derived from human NOX2/CYBB/gp91phox (142-204AA). | |
| Primary | |
| -20°C | |
| Cybb |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title