missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
Brand: Novus Biologicals NBP1-31546
This item is not returnable.
View return policy
Description
NOX1 Polyclonal specifically detects NOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| NOX1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
| EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1 | |
| Rabbit | |
| 65 kDa | |
| 100 μL | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9Y5S8 | |
| NOX1 | |
| Recombinant protein encompassing a sequence within the center region of human NOX1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 27035 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction