missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PBS Tablets
Shop All Bio Techne Products
Click to view available options
Quantity:
100 Tablets
Unit Size:
100 tablets
Description
Dilutions
Applications: Western Blot, Immunocytochemistry/ Immunofluorescence
- Western Blot
- Immunocytochemistry/ Immunofluorescence
- Immunohistochemistry 1:10-1:500
- Immunohistochemistry- Frozen
- Immunohistochemistry- Paraffin
Applications: Western Blot, Immunocytochemistry/ Immunofluorescence
Recommended Storage
Store at room temperature
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Quantity | 100 Tablets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction