missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ YB1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-324 of Human YB1 The Recombinant Human YB1 Protein is derived from E. coli. The Recombinant Human YB1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. Phosphate Buffered Saline (pH 7.4) containing 20% glycerol |
| Molecular Weight (g/mol) | 38.3kDa |
| Quantity | 0.1 mg |
| Source | Human |
| Immunogen | 1-324 Aminoacids Sequences of amino acids: MGSSHHHHHH SSGLVPRGSH MGSMSSEAET QQPPAAPPAA PALSAADTKP GTTGSGAGSG GPGGLTSAAP AGGDKKVIAT KVLGTVKWFN VRNGYGFINR NDTKEDVFVH QTAIKKNNPR KYLRSVGDGE TVEFDVVEGE KGAEAANVTG PGGVPVQGSK YAADRNHYRR YPRRRGPPRN YQQNYQNSES GEKNEGSESA PEGQAQQRRP YRRRRFPPYY MRRPYGRRPQ YSNPPVQGEV MEGADNQGAG EQGRPVRQNM YRGYRPRFRR GPPRQRQPRE DGNEEDKENQ GDETQGQQPP QRRYRRNFNY RRRRPENPKP QDGKETKAAD PPAENSSAPE AEQGGAE |
| Storage Requirements | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | YB1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction