missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ WDR5 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-334 of Human WDR5 The Recombinant Human WDR5 Protein is derived from E. coli. The Recombinant Human WDR5 Protein has been validated for the following applications: SDS-Page.
Spécification
Spécification
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl. |
| Molecular Weight (g/mol) | 38.8kDa |
| Purification Method | SDS-PAGE |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | WDR5 |
| Conjugate | Unconjugated |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu