missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VAChT/SLC18A3 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
The VAChT/SLC18A3 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 6572 |
| Species | Human |
| Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Formulation | Soluble in PBS and Water |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-91348. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter |
| Gene Symbol | SLC18A3 |
| Product Type | VAChT/SLC18A3 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction