missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TSLP Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A blocking peptide containing 19 amino acids near the center of human TSLP. Source: Synthetic (Accession #: NP_149024) The TSLP Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 85480 |
| Species | Human |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-55234. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | thymic stromal lymphopoietin |
| Gene Symbol | TSLP |
| Product Type | TSLP |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction