missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Syndecan-4 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
An un-tagged recombinant protein corresponding to the amino acids 19-145 of Human Syndecan-4 The Recombinant Human Syndecan-4 Protein is derived from E. coli. The Recombinant Human Syndecan-4 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol |
| Molecular Weight (g/mol) | 13.9kDa |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | Syndecan-4 |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >85% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction