missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Shwachman Bodian-Diamond syndrome Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.50mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-250 of Human Shwachman Bodian-Diamond syndrome The Recombinant Human Shwachman Bodian-Diamond syndrome Protein is derived from E. coli. The Recombinant Human Shwachman Bodian-Diamond syndrome Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT, 50mM NaCl, 0.1mM EDTA |
| Gene ID (Entrez) | 51119 |
| Molecular Weight (g/mol) | 30.9kDa |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Source | Human |
| Immunogen | SBDS, 1-250aa. Sequence: MGSSHHHHHHSSGLVPRGSHMSIFTPTNQIRLTNVAVVRMKRAGKRFEIACYKNKVVGWRSGVEKDLDEVLQTHSVFVNVSKGQVAKKEDLISAFGTDDQTEICKQILTKGEVQVSDKERHTQLEQMFRDIATIVADKCVNPETKRPYTVILIERAMKDIHYSVKTNKSTKQQALEVIKQLKEKMKIERAHMRLRFILPVNEGKKLKEKLKPLIKVIESEDYGQQLEIVCLIDPGCFREIDELIKKETKGKGSLEVLNLKDVEEGDEKFE |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction