missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SESN3 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
Description
A peptide corresponding to amino acids 1 - 60 of human SESN3. The SESN3 Blocking Peptide is derived from Synthetic. The SESN3 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 143686 |
| Species | Human |
| Purification Method | Affinity purified |
| Purity | >85%, by HPLC |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-44985. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | SEST3MGC29667, sestrin 3, sestrin-3 |
| Gene Symbol | SESN3 |
| Product Type | SESN3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction