missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Retinol Binding Protein RBP Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-197 of Human Retinol Binding Protein RBP The Recombinant Human Retinol Binding Protein RBP Protein is derived from E. coli. The Recombinant Human Retinol Binding Protein RBP Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_002890 |
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 200mM NaCl |
| Gene ID (Entrez) | 5947 |
| Purification Method | Protein |
| Quantity | 250 μg |
| Source | Recombinant human RBP1 protein, fused to His-tag at N-terminus, expressed in E. coli |
| Immunogen | RBP1, 1-197aa, Human, His tag, E. coli (NP_002890). |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction