missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RelA/NFkB p65 [p Ser529] Antibody Blocking Peptide

Product Code. 18018226 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18018226 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18018226 Supplier Novus Biologicals™ Supplier No. NBP197000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from human RelA/NFkB p65. Source: Synthetic The RelA/NFkB p65 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5970
Species Human
Purity >95%
Accession Number Q04206, Q04206, Q04206, Q04206
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
For Use With (Application) Control Peptide Should be Used at 1.0 μg / 1.0 μL of Antiserum in / Assay.
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Gene Symbol RELA
Product Type RelA/NFkB p65 Peptide
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.